missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) acetate (47% Pd), MilliporeSigma™

Catalog No. p-5547640
Change view
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M8180568500 500 mg Glass Bottle
M8180560005 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M8180568500 Supplier Technidata Supplier No. 8.18056.8500
Only null left

For synthesis

Chemical Identifiers

CAS 3375-31-3
Molecular Formula C4H6O4Pd
Molecular Weight (g/mol) 224.51
MDL Number MFCD00012453
InChI Key YJVFFLUZDVXJQI-UHFFFAOYSA-L
Synonym palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate
PubChem CID 167845
IUPAC Name palladium(2+) diacetate
SMILES [Pd++].CC([O-])=O.CC([O-])=O

Specifications

Quantity 500 mg
Solubility Information Soluble in water at 20°C
Formula Weight 224.5
Odor Slight
Packaging Glass Bottle
Chemical Name or Material Palladium(II) acetate (47% Pd)
Hazard Category H318: Causes serious eye damage.
EINECSNumber 222-164-4