missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium(II) acetate (47% Pd), MilliporeSigma™
For synthesis
Supplier: Technidata 8180560005
Specifications
| Palladium(II) acetate (47% Pd) | |
| 5 g | |
| MFCD00012453 | |
| Soluble in water at 20°C | |
| [Pd++].CC([O-])=O.CC([O-])=O | |
| 224.51 | |
| 224.5 | |
| Glass Bottle |
| 3375-31-3 | |
| C4H6O4Pd | |
| palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate | |
| YJVFFLUZDVXJQI-UHFFFAOYSA-L | |
| palladium(2+) diacetate | |
| 167845 | |
| Slight |
Chemical Identifiers
| 3375-31-3 | |
| 224.51 | |
| YJVFFLUZDVXJQI-UHFFFAOYSA-L | |
| 167845 | |
| [Pd++].CC([O-])=O.CC([O-])=O |
| C4H6O4Pd | |
| MFCD00012453 | |
| palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate | |
| palladium(2+) diacetate |
Safety and Handling
H318: Causes serious eye damage.
EINECSNumber : 222-164-4