missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium hydroxide on carbon, powder, unreduced, 20% Pd, moisture 50%
CAS: 12135-22-7 | H2O2Pd | 140.43 g/mol
$2,351.62 - $2,351.62
Chemical Identifiers
| CAS | 12135-22-7 |
|---|---|
| Molecular Formula | H2O2Pd |
| Molecular Weight (g/mol) | 140.43 |
| MDL Number | MFCD00064599 |
| InChI Key | NXJCBFBQEVOTOW-UHFFFAOYSA-L |
| Synonym | palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon |
| PubChem CID | 9942167 |
| SMILES | [OH-].[OH-].[Pd++] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC199620100
|
Thermo Scientific Chemicals
199620100 |
10 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12135-22-7 | |
| 140.43 | |
| NXJCBFBQEVOTOW-UHFFFAOYSA-L | |
| 9942167 |
| H2O2Pd | |
| MFCD00064599 | |
| palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon | |
| [OH-].[OH-].[Pd++] |
Specifications
| Black | |
| Powder | |
| H2O2Pd | |
| MFCD00064599 | |
| 15, 7166 | |
| Solubility in water: insoluble | |
| [OH-].[OH-].[Pd++] | |
| 140.43 | |
| 140.43 | |
| Glass bottle |
| 12135-22-7 | |
| 10 g | |
| Pd(OH)2 | |
| 16,269 | |
| palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon | |
| NXJCBFBQEVOTOW-UHFFFAOYSA-L | |
| palladium(2+);dihydroxide | |
| 9942167 | |
| 19 to 21% (Pd, on dry basis) | |
| Palladium hydroxide on carbon, 20% Pd, moisture ca 60% |
Safety and Handling
EINECSNumber : 235-219-2
TSCA : TSCA
RUO – Research Use Only