missing translation for 'onlineSavingsMsg'
Learn More

Palladium hydroxide on carbon, powder, unreduced, 20% Pd, moisture 50%

CAS: 12135-22-7 | H2O2Pd | 140.43 g/mol

Supplier:  Thermo Scientific Chemicals 199620100

 View more versions of this product

Catalog No. AC199620100

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Palladium hydroxide on carbon
Black
Powder
H2O2Pd
MFCD00064599
15, 7166
Solubility in water: insoluble
[OH-].[OH-].[Pd++]
140.43
140.43
Glass bottle
20% Pd, moisture ca 60%
12135-22-7
10 g
Pd(OH)2
16,269
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
NXJCBFBQEVOTOW-UHFFFAOYSA-L
palladium(2+);dihydroxide
9942167
19 to 21% (Pd, on dry basis)

Chemical Identifiers

12135-22-7
140.43
NXJCBFBQEVOTOW-UHFFFAOYSA-L
9942167
H2O2Pd
MFCD00064599
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
[OH-].[OH-].[Pd++]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 235-219-2

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only