missing translation for 'onlineSavingsMsg'
Learn More

Palladium, AAS standard solution, Specpure™ Pd 1000μg/mL, Thermo Scientific Chemicals

$143.15 - $143.15

Chemical Identifiers

MDL Number MFCD03613603
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA88085AE
View Documents
Thermo Scientific Chemicals
088085AE
100 mL
Each for $143.15
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD03613603

Specifications

Liquid
MFCD03613603
100 mL
Palladium, AAS standard solution, Pd 1000μg/mL
Pd in 20% HCl
UN1789
Specpure
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID

EINECSNumber : 231-595-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only