missing translation for 'onlineSavingsMsg'
Learn More

Palladium, 0.12%, Ruthenium, 0.12%; on 3mm alumina tablets, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 4220618

Quantity 50g

 View more versions of this product

Catalog No. AA4220618

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Reduction of nitrogen oxides to N2 with H2 in the presence of CO and CO2
Specifications

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03613602
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]

Specifications

Palladium
50g
MFCD03613602
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
0.12%, Ruthenium, 0.12%; on 3mm alumin.a tablets
7440-05-3
Pd
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
[Pd]
106.42
CHEBI:33363
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents