missing translation for 'onlineSavingsMsg'
Learn More

Paliperidone 97.0+%, TCI America™

Catalog No. p-7135310
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1897200MG 200 mg
P18971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1897200MG Supplier TCI America Supplier No. P1897200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 144598-75-4
Molecular Formula C23H27FN4O3
Molecular Weight (g/mol) 426.492
InChI Key PMXMIIMHBWHSKN-UHFFFAOYSA-N
Synonym paliperidone, 9-hydroxyrisperidone, invega, 9-oh-risperidone, invega sustenna, xeplion, palliperidone, 3-2-4-6-fluoro-1,2-benzisoxazol-3-yl-1-piperidinyl ethyl-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-4h-pyrido 1,2-a pyrimidin-4-one, chembl1621, 3-2-4-6-fluorobenzo d isoxazol-3-yl piperidin-1-yl ethyl-9-hydroxy-2-methyl-6,7,8,9-tetrahydro-4h-pyrido 1,2-a pyrimidin-4-one
PubChem CID 115237
ChEBI CHEBI:83804
IUPAC Name 3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-9-hydroxy-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
SMILES CC1=C(C(=O)N2CCCC(C2=N1)O)CCN3CCC(CC3)C4=NOC5=C4C=CC(=C5)F

Specifications

Color Yellow
Quantity 200 mg
UN Number 2811
Formula Weight 426.49
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Paliperidone
RTECSNumber UV1164720
TSCA No