missing translation for 'onlineSavingsMsg'
Learn More

Palipéridone 97,0+%, TCI America™

Catalog No. P1897200MG
Change view
Click to view available options
Quantité:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
P1897200MG 200 mg
P18971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1897200MG Supplier TCI America Supplier No. P1897200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 144598-75-4
Formule moléculaire C23H27FN4O3
Poids moléculaire (g/mol) 426.492
Clé InChI PMXMIIMHBWHSKN-UHFFFAOYSA-N
Synonyme paliperidone, 9-hydroxyrisperidone, invega, 9-oh-risperidone, invega sustenna, xeplion, palliperidone, 3-2-4-6-fluoro-1,2-benzisoxazol-3-yl-1-piperidinyl ethyl-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-4h-pyrido 1,2-a pyrimidin-4-one, chembl1621, 3-2-4-6-fluorobenzo d isoxazol-3-yl piperidin-1-yl ethyl-9-hydroxy-2-methyl-6,7,8,9-tetrahydro-4h-pyrido 1,2-a pyrimidin-4-one
PubChem CID 115237
ChEBI CHEBI:83804
Nom de l’IUPAC 3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)pipéridine-1-yl]éthyle]-9-hydroxy-2-méthyl-6,7,8,9-tétrahydropyrido[1,2-a]pyrimidine-4-one
SOURIRES CC1=C(C(=O)N2CCCC(C2=N1)O)CCN3CCC(CC3)C4=NOC5=C4C=CC(=C5)F

Specifications

Nom ou substance chimique Paliperidone
Couleur Jaune
Quantité 200 mg
Numéro UN 2811
Poids de formule 426.49
Pourcentage de pureté ≥97,0% (T)
Forme physique Poudre cristalline
RTECSNumber UV1164720
TSCA Non