missing translation for 'onlineSavingsMsg'
Learn More

Palatinose Hydrate 98.0+%, TCI America™

Catalog No. p-7135021
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P123425G 25 g
P1234500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P123425G Supplier TCI America Supplier No. P123425G
Only null left

Chemical Identifiers

CAS 343336-76-5
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD11100373
InChI Key PVXPPJIGRGXGCY-VVLSOAGWNA-N
Synonym 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4s,5r-3,4,5-trihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl methoxy tetrahydro-2h-pyran-3,4,5-triol hydrate
PubChem CID 126970043
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4S,5S)-3,4,5-trihydroxy-5-(hydroxymethyl)oxolan-2-yl]methoxy}oxane-3,4,5-triol
SMILES OC[C@H]1O[C@H](OC[C@H]2O[C@@](O)(CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 123°C
Color White
Quantity 25 g
Formula Weight 342.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Palatinose Hydrate