missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PAK7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115717
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115717 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115717 Supplier Invitrogen™ Supplier No. PA5115717
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PAK7.

PAK7 (KIAA1264) is implicated in the regulation of cytoskeletal dynamics, proliferation, and cell survival signaling. This kinase contains a CDC42/Rac1 interactive binding motif, and has been shown to bind CDC42 in the presence of GTP. This kinase is predominantly expressed in brain. It is capable of promoting neurite outgrowth, and thus may play a role in neurite development. This kinase is associated with microtubule networks and induces microtubule stabilization. The subcellular localization of this kinase is tightly regulated during cell cycle progression. Alternatively spliced transcript variants encoding the same protein have been described.

Specifications

Antigen PAK7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene PAK5
Gene Accession No. D4A280, Q8C015, Q9P286
Gene Alias 2900083L08Rik; 6430627N20; KIAA1264; p21 (CDKN1A)-activated kinase 7; p21 (RAC1) activated kinase 5; p21 (RAC1) activated kinase 7; p21 protein (Cdc42/Rac)-activated kinase 7; p21(CDKN1A)-activated kinase 7; p21-activated kinase 5; p21-activated kinase 7; p21CDKN1A-activated kinase 7; PAK 5; PAK 7; PAK5; PAK-5; Pak7; PAK-7; protein kinase PAK5; RP5-1119D9.3; Serine/threonine-protein kinase PAK 5; serine/threonine-protein kinase PAK 7; serine/threonine-protein kinase PAK7
Gene Symbols PAK5, Pak7
Host Species Rabbit
Immunogen A synthesized peptide derived from human PAK5(Accession Q9P286), corresponding to amino acid residues S286-A336.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 241656, 311450, 57144
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less