missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAH Monoclonal Antibody (C4), Invitrogen™

Description
PAH encodes the enzyme phenylalanine hydroxylase that is the rate-limiting step in phenylalanine catabolism. Deficiency of this enzyme activity results in the autosomal recessive disorder phenylketonuria.
Specifications
Specifications
| Antigen | PAH |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | C4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 |
| Gene | PAH |
| Gene Accession No. | P00439, P04176, P16331 |
| Gene Alias | AW106920; Pah; PH; phe-4-monooxygenase; phenylalanine 4-monooxygenase; phenylalanine hydroxylase; phenylalanine-4-hydroxylase; PKU; PKU1 |
| Show More |