missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polynuclear Aromatic Hydrocarbons Combination Mix, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC 8310A
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Acenaphthene | 83-32-9 | 500μg/mL |
| Acenaphthylene | 208-96-8 | 500μg/mL |
| Anthracene | 120-12-7 | 500μg/mL |
| Benz(a)anthracene | 56-55-3 | 500μg/mL |
| Benzo(a)pyrene | 50-32-8 | 500μg/mL |
| Benzo(b)fluoranthene | 205-99-2 | 500μg/mL |
| Benzo(g,h,i)perylene | 191-24-2 | 500μg/mL |
| Benzo(k)fluoranthene | 207-08-9 | 500μg/mL |
| Chrysene | 218-01-9 | 500μg/mL |
| Dibenz(a,h)anthracene | 53-70-3 | 500μg/mL |
| Fluoranthene | 206-44-0 | 500μg/mL |
| Fluorene | 86-73-7 | 500μg/mL |
| Indeno(1,2,3-c,d)pyrene | 193-39-5 | 500μg/mL |
| Naphthalene | 91-20-3 | 500μg/mL |
| Phenanthrene | 85-01-8 | 500μg/mL |
| Pyrene | 129-00-0 | 500μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| PAH Combination Mix | |
| 2mL amber prescored glass ampule | |
| For US EPA Methods 8151, 8310 | |
| GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
| Acetonitrile |
| SW-846 Method 8310 | |
| Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. | |
| AA, ICP | |
| Certified Reference Material | |
| 1 mL |
Product intended for laboratory use only