missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAGE3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104046
Description
Antibody detects endogenous levels of total PAGE3.
PAGE-3 (prostate-associated gene 3 protein), also known as GAGED1 (putative G antigen family Dmember 1), is a 113 amino acid protein that belongsto the GAGE family. The gene that encodes PAGE-3 consists of approximately 6,432 bases andmapsto human chromosome Xp11.21. Chromosome X consists of about 153million base pairs and nearly 1,000 genes. The combination of a X and Y chromosome lead to normal male development while two copies of X lead to normal female development. There are a number ofconditionsrelated to an unusual number and combination ofsexchromosomes being inherited, including Turner's syndrome, Klinefelter'ssyndrome and Triple X syndrome. Color blindness, hemophilia, and Duchennemuscular dystrophy arewellknown X chromosome-linked conditionswhich affectmalesmore frequently as males carry a single X chromosome.Chemical Identifiers
| PAGE3 | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| PAGE3 | |
| CT16.6; G antigen family D member 1; G antigen, family D, 1; GAGED1; P antigen family member 3; P antigen family, member 3 (prostate associated); PAGE family member 3; PAGE3; PAGE-3; Prostate-associated gene 3 protein | |
| PAGE3 | |
| Primary | |
| Affinity Chromatography |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5JUK9 | |
| 139793 | |
| A synthesized peptide derived from human PAGE3(Accession Q5JUK9), corresponding to amino acid residues G25-L75. | |
| Antibody |
Specifications
| PAGE3 | |
| Polyclonal | |
| Unconjugated | |
| PAGE3 | |
| CT16.6; G antigen family D member 1; G antigen, family D, 1; GAGED1; P antigen family member 3; P antigen family, member 3 (prostate associated); PAGE family member 3; PAGE3; PAGE-3; Prostate-associated gene 3 protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 139793 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5JUK9 | |
| PAGE3 | |
| A synthesized peptide derived from human PAGE3(Accession Q5JUK9), corresponding to amino acid residues G25-L75. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |