missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PADI6 Monoclonal Antibody (D2)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA566096
Description
PADI6 Monoclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes a member of the peptidyl arginine deiminase family of enzymes, which catalyze the post-translational deimination of proteins by converting arginine residues into citrullines in the presence of calcium ions. The family members have distinct substrate specificities and tissue-specific expression patterns. This protein may play a role in cytoskeletal reorganization in the egg and in early embryo development.Chemical Identifiers
| PADI6 | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q6TGC4 | |
| 353238, 414431 | |
| Recombinant Peptidyl Arginine Deiminase Type VI (PADI6) (Ser393-Pro694). | |
| Antibody |
| D2 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2b κ | |
| Human, Pig | |
| PADI6 | |
| arginine deiminase-like protein; Egg and embryo abundant PAD; ePAD; hPADVI; PAD6; Padi5; Padi6; peptidyl arginine deiminase 6; peptidyl arginine deiminase, egg and embryo abundant; peptidyl arginine deiminase, type VI; peptidyl arginine deiminase-like protein; peptidylarginine deiminase type 6; peptidylarginine deiminase VI; Protein-arginine deiminase type VI; protein-arginine deiminase type-6 | |
| PADI6 | |
| Primary | |
| Protein A/G |
Specifications
| PADI6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q6TGC4 | |
| PADI6 | |
| Recombinant Peptidyl Arginine Deiminase Type VI (PADI6) (Ser393-Pro694). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| D2 | |
| Unconjugated | |
| PADI6 | |
| arginine deiminase-like protein; Egg and embryo abundant PAD; ePAD; hPADVI; PAD6; Padi5; Padi6; peptidyl arginine deiminase 6; peptidyl arginine deiminase, egg and embryo abundant; peptidyl arginine deiminase, type VI; peptidyl arginine deiminase-like protein; peptidylarginine deiminase type 6; peptidylarginine deiminase VI; Protein-arginine deiminase type VI; protein-arginine deiminase type-6 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 353238, 414431 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |