missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Pack of 25 Individually Wrapped Sterile Polypropylene Centrifuge Tubes with Flat Cap, Non-Self Standing

Pack of 25 sterile polypropylene centrifuge tubes with flat caps are individually wrapped for contamination protection, leak-resistant, and chemically durable, ideal for secure sample handling in sensitive lab and research applications.

$101.25 - $132.64

Specifications

Color Transparent
Disposable Reusable
Material Polypropylene
Sterility Sterile
Autoclavable Autoclavable
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
S000161
View Documents Promotion Details
Eisco Labs
CH5623PK25
15 mL
Each for $101.25
Only null left
 
S000162
View Documents Promotion Details
Eisco Labs
CH5624PK25
50 mL
Each for $132.64
Only null left
 
Description

Description

  • Sterile centrifuge tubes: Pack of 25 polypropylene tubes with flat caps, individually wrapped for secure storage.
  • Individually wrapped design: Ensures protection from external exposure, ideal for sensitive applications.
  • Secure flat cap: Prevents leaks and evaporation, offering reliable sample storage.
  • Versatile lab applications: Suitable for sample preparation, centrifugation, molecular biology, and general research use.
  • Durable polypropylene construction: Designed for strength and chemical resistance, ensuring reliable performance in laboratory settings.
Specifications

Specifications

Transparent
Polypropylene
Autoclavable
Conical
Yes
Yes
24,000 x g
Reusable
Sterile
Cylindrical
Flat Cap
Yes
Centrifuge Tube
Videos
SDS
Documents

Documents