missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Pack of 12 Narrow Mouth HDPE Translucent Reagent Bottles with Polypropylene Caps

Catalog No. S000082
Change view
Click to view available options
Capacity (Metric):
4 mL
15 mL
30 mL
500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000082 4 mL
S000083 15 mL
S000084 30 mL
S000085 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S000082 Supplier Eisco Labs Supplier No. CH5361PK12

Narrow mouth HDPE reagent bottles with secure polypropylene caps that are leak-tested, chemically resistant, and ideal for safe storage and transfer of lab liquids in clean, high-performance environments.

  • HDPE Reagent Bottles: Includes 12 narrow mouth translucent HDPE bottles with polypropylene caps. Designed for safe and effective storage in labs or industrial environments.
  • Sterile With Chemical Resistance: Manufactured sterile for clean applications. HDPE construction offers excellent resistance to a variety of chemicals, making it ideal for reagent storage and transport.
  • Leak-Proof Tested For Safety: Each bottle is rigorously tested to ensure a reliable seal and leak-proof performance. Filled and inverted under pressure with no liquid escape.
  • Designed For Lab Use: These bottles allow easy access for filling or pouring. The translucent material enables liquid visibility for efficient handling and monitoring.
  • Engineered For Reliability: Produced in a Class 100K clean room, these bottles meet stringent standards for lab and industrial use. Built for durability with a temperature range of -100°C to +120°C.

Specifications

Capacity (Metric) 4 mL
Capacity (English) 0.13 oz.
Color Translucent
Diameter (Metric) Inner Neck 8.3 mm
Material HDPE
No. per Pack 12 Pk.
Sterility Sterile
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 41 mm
Product Type Reagent Bottle
Diameter (Metric) 16 mm
Autoclavable Not Autoclavable
Show More Show Less