missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Pacific Orange™ and Pacific Blue™ Antibody Labeling Kits

Catalog No. p-7227925
Encompass
Change view
Click to view available options
Excitation/Emission:
405/551 nm
410/455 nm
Color:
Pacific Blue
Pacific Orange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Excitation/Emission Color
501121574 405/551 nm Pacific Orange
P30013 410/455 nm Pacific Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 501121574 Supplier Molecular Probes™ Supplier No. P30014
Only null left

Antibody Labeling Kits specifically bind small amounts of antibody with violet light-excitable Pacific Orange or Pacific Blue dyes. Conveniently label up to 100 μg of antibody per reaction, with 5 reactions per kit.

Pacific Orange and Pacific Blue antibody labeling kits enable attachment of a violet light-excitable fluorescent label on antibodies (up to 100 μg per reaction). Each kit contains enough dye for five reactions.

A primary antibody directly labeled with a fluorophore often produces lower background fluorescence and less nonspecific binding. Primary antibodies of the same isotype or derived from the same species can be multiplexed if they are directly labeled with compatible fluorophores. Using Pacific Blue and Pacific Orange dyes simultaneously allows you to excite both dyes at the same wavelength while their emissions are differentially distinguished.

Pacific Orange and Pacific Blue antibody labeling kits allow the labeling of small amounts of antibody with a violet light-excitable dye (up to 100 μg of antibody per reaction). Small amounts of other proteins (>40 kDa) can also be labeled using these kits.

Features of these antibody labeling kits
• Pacific Blue excitation and emission maxima: 410/455 nm
• Pacific Orange excitation and emission maxima: 405/551 nm
• Labeled proteins typically ready to use in 90 min. (∼15 min. hands-on time)
• Optimized for small-scale labeling of any protein >40 kDa
• Stabilizing proteins should be removed from the sample before labeling
• Purified using convenient spin filters
• Includes detailed instructions for determining degree of labeling (DOL)

These kits contain everything you need to perform five separate labeling reactions as well as to purify the resulting conjugates. Conjugates are ideal for multiple applications, including flow cytometry, fluorescent microscopy, immunohistochemistry, primary detection, ELISA, immunocytochemistry, indirect FISH, and more. We are ISO 13485:2000 certified.

Specifications

Product Type Antibody Labeling Kit
Quantity 5 Reactions kit
Content And Storage Pacific Orange™ Antibody Labeling Kit comes with 5 vials of reactive dye, sodium bicarbonate, purification resin, spin columns and collections tubes.

Store at 2-8°C, protected from light. Component A may be stored from -20 to 8°C.
Detection Method Fluorescence
Excitation/Emission 405/551 nm
Labeling Scale 100 μg
Shipping Condition Room Temperature
Product Line Pacific Orange
Chemical Reactivity Amine
Solubility DMSO (Dimethylsulfoxide)
Label or Dye Pacific Orange
Color Pacific Orange
Labeling Target Antibodies, Proteins
Label Type Pacific Dyes
Show More Show Less