Learn More
Thermo Scientific™ Pacific Hemostasis™ Factor Assays
Fisherbrand™ Pacific Hemostasis Factor Assays are used for the detection and quantification of factor deficiencies in patient plasmas.
$624.35
Description
Detect and quantify factor deficiencies in patient plasmas with Thermo Scientific™ Pacific Hemostasis™ Factor Assays.
Pacific Hemostasis Factor Assays include a complete line of lyophilized congenital factor deficient plasmas for performing factor assays.- All factor deficient plasmas are prepared from congenitally deficient patient donors or immunodepleted plasma, with the exception of Factor II.
- Factor II Deficient Plasma is a mix of human and bovine plasma.
- Plasmas are packaged 10 × 1mL; Diluting Fluid is packaged 2 × 100mL.