missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pacific Blue™ Succinimidyl Ester
Description
The amine-reactive Pacific Blue™ succinimidyl ester can be used to can be used to create blue-fluorescent bioconjugates with excitation/emission maxima ∼410/455 nm that are excitable by the 405 nm spectral line of the blue diode (violet) laser.
Related Links:
- View all Pacific Blue™ dye products..
- View the Fluorophore Selection Guide.
Specifications
Specifications
| Format | Solid |
| Quantity | 5 mg |
| Product Type | Succinimidyl Ester |
| Solubility | DMSO (Dimethylsulfoxide) |
| Content And Storage | Contains 1 vial (1 mg). Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Detection Method | Fluorescence |
| Flow Cytometer Laser Lines | 405 |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Active Ester, Succinimidyl Ester |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.