Learn More
Thermo Scientific™ μPAC™ Neo Plus ESI-MS Accessories
Catalog No. 01258268
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Type:
Emitter
Interface
Length (Metric):
N/A
Diameter (Metric) Inner:
N/A
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Length (Metric) Diameter (Metric) Inner
01258268 Emitter N/A N/A
01258269 Interface N/A N/A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. 01258268 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' EMIEASYSPRAYB
Use μPAC™ ESI-MS Accessories for fast, reproducible, and low dispersion grounded connection between Thermo Scientific™ μPAC™ Neo Plus HPLC columns and Thermo Scientific™ Nanospray Flex™ and EASY-Spray™ ion sources.
Use Thermo Scientific™ μPAC™ ESI-MS Accessories to facilitate reproducible, low dispersion, and grounded connections between μPAC Neo Plus HPLC columns and low-flow mass spectrometer ion sources. μPAC Neo Plus voltage spacers are available for both EASY-Spray and Nanospray Flex ion sources. All configurations require a grounding cable.
The μPAC Flex iON Connect ESI-MS Interface consists of two separate units:
  • A docking unit that can be slid onto the Nanospray Flex Ion Source's metal rod and receives both a grounding and a high voltage cable
  • A spray unit compatible with the outlet fittings of μPAC columns on one side and compatible with any 360 μm OD emitter on the other side.
The μPAC EASY-Spray emitter is designed for easy connection to an EASY-Spray MS Source. It contains a pre-assembled EASY-Spray emitter with a transfer line and grounding union.
Specifications
Description μPAC™ Compatible EASY-Spray Emitter
For Use With (Equipment) EASY-Spray™ Source
Type Emitter
Length (Metric) N/A
Diameter (Metric) Inner N/A