Learn More
Motic™ PA53 MET Trinocular Upright Materials Microscope
A combination of technology and creativity, the PA53 MET series offers superior optical performance as well as an advanced ergonomic design, futuristic digital capabilities, and innovative modules ideal for metallurgy.
Supplier: Motic™ 1100104600861
Description
- Scalable Infinity-Corrected Optical System (CCIS™)
- Superior optical performance with newly designed Semi-Apochromatic objectives
- Interference Contrast (DIC) concept incident illumination
- Established contrast methods
- Multiple degrees of motorization in X, Y, and Z
- Harmonized Brightfield/Darkfield switch
- Transmitted Light: LB daylight filter (Halogen lamp)
- Diopter adjustable
Specifications
| 10X | |
| PA53 MET | |
| Reflected and Transmitted Illumination |
| S-APO Plan 5X, 10X, 20X, 50X | |
| Trinocular | |
| WF 10X/25 mm |