Learn More
Motic™ PA43 BIO Trinocular Routine Laboratory Microscopes
Description
- Scalable Infinity-Corrected Optical System (CCIS™)
- Full power transmitted Köhler illumination
- Phase contrast methods for various microbiology applications
- Darkfield accessories for blood cell studies for natural therapy
- Polarizing contrast accessories for protein crystal, tissue, gout and kidney stone observations
- Environmentally friendly 'ECO' function
- Consistent, even illumination with IL Function
- Rackless stage
- Diopter Adjustment: ±5
Specifications
Specifications
| Magnification Power | 10X |
| Objective Types | UC Plan 4X and UC PH Plan 10X, 40X, 100X |
| Head Style | Trinocular |
| Lighting Type | Kohler Illumination |
| Eyepiece | WF 10X/22 mm |