Learn More
Motic™ PA43 BIO Trinocular Routine Laboratory Microscopes
The PA43 BIOMED combines modularity with ergonomics to improve application imaging workflows. A progression into routine work, this microscope’s robust design and solid footprint does not compromise on optical quality and features.
Supplier: Motic™ 1100105500021
Description
- Scalable Infinity-Corrected Optical System (CCIS™)
- Full power transmitted Köhler illumination
- Phase contrast methods for various microbiology applications
- Darkfield accessories for blood cell studies for natural therapy
- Polarizing contrast accessories for protein crystal, tissue, gout and kidney stone observations
- Environmentally friendly 'ECO' function
- Consistent, even illumination with IL Function
- Rackless stage
- Diopter Adjustment: ±5
Chemical Identifiers
| 10X | |
| Trinocular | |
| Kohler Illumination | |
| WF 10X/22 mm |
Specifications
| 10X | |
| Trinocular | |
| WF 10X/22 mm |
| UC Plan 4X, 100X and UC PH Plan 10X, 40X | |
| Kohler Illumination |