Learn More
Motic™ PA43 BIO Trinocular Ergonomic Upright Pathology Lab Microscope
Description
- Scalable Infinity-Corrected Optical System (CCIS™)
- Full power transmitted Köhler illumination
- Tilting trinocular tube head with inverted image
- Achromat swing-out condenser
- Environmentally friendly 'ECO' function
- Consistent, even Illumination with IL function
- Rackless stage
- Diopter Adjustable: ±5
Specifications
Specifications
| Magnification Power | 10X |
| Objective Types | UC Plan 2X, 4X, 10X, 20X, 40X |
| Head Style | Trinocular |
| Lighting Type | LED, Halogen |
| Eyepiece | WF 10X/22 mm |