Learn More
Motic™ PA43 BIO Trinocular Ergonomic Pathology Lab Microscope Package
Description
- Scalable Infinity-Corrected Optical System (CCIS™)
- Full power transmitted Köhler illumination
- Tilting trinocular rube head with inverted image
- Achromat Swing-out Condenser
- Environmentally friendly 'ECO' function
- Consistent, even illumination with IL Function
- Rackless stage
- Diopter Adjustment: ±5
Specifications
Specifications
| Magnification Power | 10X |
| Objective Types | UC Plan 2X, 4X, 10X, 20X, 40X |
| Head Style | Trinocular |
| Camera Type | 6 MP Digital |
| Lighting Type | LED, Halogen |
| Eyepiece | WF 10X/22 mm |