missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p70 S6 Kinase Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528597
Description
Recommended positive controls: 293T, A431, Jurkat, Raji, HeLa. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (91%), Rabbit (100%), Chicken (96%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RPS6KB1 (p70S6K) is a serine/threonine kinase with 2 non-identical kinase catalytic domains which phosphorylates several residues of the S6 ribosomal protein. p70 S6 kinase is predominantly localized in the cytoplasm, and is essential in growth factors regulated cell proliferation, pathways involving cell motility, such as metastases, the immune response, and tissue repair. p70 S6 kinase acts downstream of phosphoinositide (PI) 3-kinase, and its main physiological target is the S6 ribosomal protein, which is involved in upregulation of protein synthesis. The kinase activity of p70 S6 kinase leads to an increase in protein synthesis and cell proliferation. Amplification of the region of DNA encoding p70 S6 kinase and overexpression are seen in some breast cancer cell lines.Chemical Identifiers
| p70 S6 Kinase | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| RPS6KB1 | |
| 10539; 2610318I15Rik; 4732464A07Rik; 70 kDa ribosomal protein S6 kinase 1; 7084; 70kDa; 7-10; AA959758; AI256796; AI314060; CG10539; CG10539-PA; CG10539-PB; Dmel\CG10539; Dmel_CG10539; Dp70[s6k]; Dp70S6k; dps6k; dS6 kinase; dS6K; d-S6K; dS6K1; EC 2.7.11.1; fs(3)07084; hypothetical protein; kinase p70S6K; KS6B1; l(3)07084; p70 p70-S6K; p70 ribosomal S6 kinase; p70 ribosomal S6 kinase alpha; p70 S6 kinase; p70 S6 kinase 1; p70 S6 kinase alpha; p70 S6 kinase, alpha; p70 S6K; p70 S6KA; p70 S6K-alpha; p70(S6K)-alpha; p70/85s6k; p70/S6K; p70[S6 kinase]; p70[S6k]; p70[S6kinase]; p70-alpha; p70S6 kinase; p70S6k; p70-S6K; p70-S6K 1; p70S6K kinase; p70s6k protein kinase homolog; P70S6K1; p70S6K-alpha; p80-S6K 1; p85 p85S6K; Pk64F; PkS6; protein kinase-like 64F; PS6K; p-S6K; ribosomal protein S6 kinase; ribosomal protein S6 kinase B1; ribosomal protein S6 kinase beta-1; Ribosomal protein S6 kinase I; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 70kD, polypeptide 1; ribosomal protein S6 kinase, 70kDa, polypeptide 1; ribosomal protein S6 kinase, polypeptide 1; ribosomal S6 protein kinase; Rps6kb1; RPS6-p70-protein kinase; S6 K; S6 kinase; s6k; S6K kinase; S6K1; s6k11; S6K-beta-1; S6kinase; S6-kinase; S6k-PA; S6k-PB; serine/threonine kinase 14 alpha; serine/threonine-protein kinase 14A; STK14A | |
| RPS6KB1 | |
| Primary | |
| Antigen affinity chromatography |
| 0.7 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P23443, P67999, Q8BSK8 | |
| 6198, 72508, 83840 | |
| Recombinant protein encompassing a sequence within the center region of human p70 S6K. The exact sequence is proprietary. | |
| Antibody |
Specifications
| p70 S6 Kinase | |
| Polyclonal | |
| Unconjugated | |
| RPS6KB1 | |
| 10539; 2610318I15Rik; 4732464A07Rik; 70 kDa ribosomal protein S6 kinase 1; 7084; 70kDa; 7-10; AA959758; AI256796; AI314060; CG10539; CG10539-PA; CG10539-PB; Dmel\CG10539; Dmel_CG10539; Dp70[s6k]; Dp70S6k; dps6k; dS6 kinase; dS6K; d-S6K; dS6K1; EC 2.7.11.1; fs(3)07084; hypothetical protein; kinase p70S6K; KS6B1; l(3)07084; p70 p70-S6K; p70 ribosomal S6 kinase; p70 ribosomal S6 kinase alpha; p70 S6 kinase; p70 S6 kinase 1; p70 S6 kinase alpha; p70 S6 kinase, alpha; p70 S6K; p70 S6KA; p70 S6K-alpha; p70(S6K)-alpha; p70/85s6k; p70/S6K; p70[S6 kinase]; p70[S6k]; p70[S6kinase]; p70-alpha; p70S6 kinase; p70S6k; p70-S6K; p70-S6K 1; p70S6K kinase; p70s6k protein kinase homolog; P70S6K1; p70S6K-alpha; p80-S6K 1; p85 p85S6K; Pk64F; PkS6; protein kinase-like 64F; PS6K; p-S6K; ribosomal protein S6 kinase; ribosomal protein S6 kinase B1; ribosomal protein S6 kinase beta-1; Ribosomal protein S6 kinase I; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 70kD, polypeptide 1; ribosomal protein S6 kinase, 70kDa, polypeptide 1; ribosomal protein S6 kinase, polypeptide 1; ribosomal S6 protein kinase; Rps6kb1; RPS6-p70-protein kinase; S6 K; S6 kinase; s6k; S6K kinase; S6K1; s6k11; S6K-beta-1; S6kinase; S6-kinase; S6k-PA; S6k-PB; serine/threonine kinase 14 alpha; serine/threonine-protein kinase 14A; STK14A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6198, 72508, 83840 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.7 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P23443, P67999, Q8BSK8 | |
| RPS6KB1 | |
| Recombinant protein encompassing a sequence within the center region of human p70 S6K. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |