missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ P2Y13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534084
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Gibbon, Marmoset (93%) Panda, Dog (87%).
The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is activated by ADP.Chemical Identifiers
| P2Y13 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| P2RY13 | |
| 2010001L06Rik; FKSG77; G protein-coupled receptor 68; G protein-coupled receptor 86; GPCR1; GPR86; GPR94; G-protein coupled receptor 86; G-protein coupled receptor 94; P2ry13; P2Y purinoceptor 13; P2Y13; P2yr13; purinergic receptor P2Y G-protein coupled 13; purinergic receptor P2Y, G-protein coupled 13; purinergic receptor P2Y, G-protein coupled, 13; purinergic receptor P2Y13; SP174; Vpurinergic receptor P2Y, G-protein coupled 13 | |
| P2RY13 | |
| Primary | |
| Antigen affinity chromatography |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.1% sodium azide | |
| Q9BPV8 | |
| 53829 | |
| Synthetic 15 amino acid peptide from N-terminal extracellular domain of human P2RY13 / P2Y13. | |
| Antibody |
Specifications
| P2Y13 | |
| Polyclonal | |
| Unconjugated | |
| P2RY13 | |
| 2010001L06Rik; FKSG77; G protein-coupled receptor 68; G protein-coupled receptor 86; GPCR1; GPR86; GPR94; G-protein coupled receptor 86; G-protein coupled receptor 94; P2ry13; P2Y purinoceptor 13; P2Y13; P2yr13; purinergic receptor P2Y G-protein coupled 13; purinergic receptor P2Y, G-protein coupled 13; purinergic receptor P2Y, G-protein coupled, 13; purinergic receptor P2Y13; SP174; Vpurinergic receptor P2Y, G-protein coupled 13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53829 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9BPV8 | |
| P2RY13 | |
| Synthetic 15 amino acid peptide from N-terminal extracellular domain of human P2RY13 / P2Y13. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |