missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ P2X2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119683
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119683 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5119683 Supplier Invitrogen™ Supplier No. PA5119683
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Positive Control: Mouse brain tissue lysates, rat brain tissue, human esophagus tissue, mouse brain tissue, MCF-7. Subcellular Location: Cell membrane.

P2rx2 are members of the ligand-gated ion channel family that open in response to extracellular ATP. Each receptor is made up of a trimer of subunits (P2X1-7) all of which share the common structure of two transmembrane domains. P2rx2 functions as a ligand-gated ion channel and binding to ATP mediates synaptic transmission between neurons and from neurons to smooth muscle. Six transcript variants encoding six distinct isoforms have been identified for P2rx2. P2rx2 is involved in a variety of cellular responses, such as excitatory postsynaptic responses in sensory neurons, neuromuscular junctions (NMJ) formation, hearing, perception of taste and peristalsis. In the inner ear, P2rx2 regulates sound transduction and auditory neurotransmission, outer hair cell electromotility, inner ear gap junctions, and K(+) recycling. P2rx2 also mediates synaptic transmission between neurons and from neurons to smooth muscle. Diseases associated with P2RX2 include Deafness, Autosomal Dominant 41 and Dfna41 Nonsyndromic Hearing Loss And Deafness.

Specifications

Antigen P2X2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene P2RX2
Gene Accession No. P49653, Q8K3P1, Q9UBL9
Gene Alias ATP receptor; DFNA41; MGC129601; P2RX2; P2X purinoceptor 2; P2X Receptor, subunit 2; P2X2; P2X2 purinoceptor; P2X2 receptor splice variant P2X2-1; P2X2 receptor splice variant P2X2-2; P2X2 receptor splice variant P2X2-3; P2X2a; Purinergic receptor; purinergic receptor P2X 2; purinergic receptor P2X, ligand gated ion channel, 2; purinergic receptor P2X, ligand-gated ion channel, 2; purinergic receptor P2X2
Gene Symbols P2RX2
Host Species Rabbit
Immunogen Synthetic peptide within C-terminal mouse P2X2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114115, 22953, 231602
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less