missing translation for 'onlineSavingsMsg'
Learn More

Dentec Safety™ P100 Low Profile Cartridge

Dentec Safety™ P100 Low Profile Cartridge is uniquely designed with a space plenum between the filtering material and the inhalation port

$18.48 - $719.85

Specifications

Color Purple
Material Plastic
Product Type Respirator Accessories
For Use With (Equipment) Masks
Includes Cartridges and filters
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
19222502
View Documents
Dentec Safety™
DTC6451832TLPOCAM
1 Pair
Pair for $18.48
Only null left
 
19222500
View Documents
Dentec Safety™
DTC6451830TLPO
6/box
Pack of 6 for $49.42
Only null left
 
19222501
View Documents
Dentec Safety™
DTC6451831TLPOB100
100/Cs.
Case of 100 for $719.85
Only null left
 
Description

Description

  • Inhaled air is evenly distributed across the filter, lowering breathing resistance and extending cartridge life.
  • Ultra thin design fits under any grinding shield or welding helmet.
  • Will not obstruct field of vision
  • Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
  • Offset connector improves visibility and allows face shields or welding helmets to be worn without interference
Specifications

Specifications

Purple
Respirator Accessories
Cartridges and filters
Plastic
Masks
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'