missing translation for 'onlineSavingsMsg'
Learn More

Dentec Safety™ P100 Low Profile Cartridge

Catalog No. 19222502
Change view
Click to view available options
Quantity:
1 Pair
100/Cs.
6/box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19222502 1 Pair
19222500 6/box
19222501 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19222502 Supplier Dentec Safety™ Supplier No. DTC6451832TLPOCAM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dentec Safety™ P100 Low Profile Cartridge is uniquely designed with a space plenum between the filtering material and the inhalation port

  • Inhaled air is evenly distributed across the filter, lowering breathing resistance and extending cartridge life.
  • Ultra thin design fits under any grinding shield or welding helmet.
  • Will not obstruct field of vision
  • Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
  • Offset connector improves visibility and allows face shields or welding helmets to be worn without interference

Specifications

Color Purple
Material Plastic
Product Type Respirator Accessories
For Use With (Equipment) Masks
Includes Cartridges and filters
Quantity 1 Pair