missing translation for 'onlineSavingsMsg'
Learn More

p-Xylylenediphosphonic Acid 97.0+%, TCI America™

Catalog No. X00745G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00745G 5 g
X00741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X00745G Supplier TCI America Supplier No. X00745G

Chemical Identifiers

CAS 4546-06-9
Molecular Formula C8H12O6P2
Molecular Weight (g/mol) 266.13
MDL Number MFCD05663703
InChI Key ZURHBENZJDSCRG-UHFFFAOYSA-N
PubChem CID 155037
IUPAC Name [4-(phosphonomethyl)phenyl]methylphosphonic acid
SMILES C1=CC(=CC=C1CP(=O)(O)O)CP(=O)(O)O

Specifications

Melting Point 280°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 266.13
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material p-Xylylenediphosphonic Acid