missing translation for 'onlineSavingsMsg'
Learn More

p-Xylylenediamine 99.0+%, TCI America™

Catalog No. p-7126559
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D101825G 25 g
D1018100G 100 g
D1018500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D101825G Supplier TCI America Supplier No. D101825G
Only null left

Chemical Identifiers

CAS 539-48-0
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00009821
InChI Key ISKQADXMHQSTHK-UHFFFAOYSA-N
Synonym p-xylylenediamine, 1,4-phenylenedimethanamine, 1,4-bis aminomethyl benzene, 1,4-benzenedimethanamine, 1,4-xylylenediamine, p-xylylendiamine, 4-aminomethylbenzylamine, p-xylylendiamine czech, p-phenylenebis methylamine, 4-aminomethyl phenyl methanamine
PubChem CID 68315
IUPAC Name [4-(aminomethyl)phenyl]methanamine
SMILES C1=CC(=CC=C1CN)CN

Specifications

Melting Point 62°C
Color White-Yellow
Boiling Point 125°C
Quantity 25 g
UN Number 3259
Formula Weight 136.20
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material p-Xylylenediamine
EINECSNumber (3)-0308
RTECSNumber PF9000000
TSCA Yes