Learn More
p-Xylylenediamine, 97%
CAS: 539-48-0 | C8H12N2 | 136.198 g/mol
Supplier: Thermo Scientific Chemicals L0299518
Description
p-Xylylenediamine is
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsp-Xylylenediamine is
Solubility
Soluble in water.
Notes
Air Sensitive. Incompatible with air, carbondioxide and oxidizing agents. Store underdry inert gas.
Specifications
| p-Xylylenediamine | |
| 59°C to 63°C | |
| 134°C (273°F) | |
| MFCD00009821 | |
| UN3259 | |
| Air Sensitive | |
| Soluble in water. | |
| C1=CC(=CC=C1CN)CN | |
| 136.198 | |
| 136.2 |
| 539-48-0 | |
| 230°C (10 mmHg) | |
| C8H12N2 | |
| 50 g | |
| 2206190 | |
| p-xylylenediamine, 1,4-phenylenedimethanamine, 1,4-bis aminomethyl benzene, 1,4-benzenedimethanamine, 1,4-xylylenediamine, p-xylylendiamine, 4-aminomethylbenzylamine, p-xylylendiamine czech, p-phenylenebis methylamine, 4-aminomethyl phenyl methanamine | |
| ISKQADXMHQSTHK-UHFFFAOYSA-N | |
| [4-(aminomethyl)phenyl]methanamine | |
| 68315 | |
| 97% |
Chemical Identifiers
| 539-48-0 | |
| 136.198 | |
| ISKQADXMHQSTHK-UHFFFAOYSA-N | |
| 68315 | |
| C1=CC(=CC=C1CN)CN |
| C8H12N2 | |
| MFCD00009821 | |
| p-xylylenediamine, 1,4-phenylenedimethanamine, 1,4-bis aminomethyl benzene, 1,4-benzenedimethanamine, 1,4-xylylenediamine, p-xylylendiamine, 4-aminomethylbenzylamine, p-xylylendiamine czech, p-phenylenebis methylamine, 4-aminomethyl phenyl methanamine | |
| [4-(aminomethyl)phenyl]methanamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber : 208-719-3
RTECSNumber : PF9000000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only