missing translation for 'onlineSavingsMsg'
Learn More

p-Xylylenediamine, 97%

Catalog No. aal0299504
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
2 g
50 g
Catalog No. Quantity
AAL0299504 2 g
AAL0299509 10 g
AAL0299518 50 g
3 options

Catalog No. AAL0299504

Supplier: Thermo Scientific Chemicals L0299504

Only null left

CAS: 539-48-0 | C8H12N2 | 136.198 g/mol

p-Xylylenediamine is

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Xylylenediamine is

Solubility
Soluble in water.

Notes
Air Sensitive. Incompatible with air, carbondioxide and oxidizing agents. Store underdry inert gas.

Chemical Identifiers

CAS 539-48-0
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00009821
InChI Key ISKQADXMHQSTHK-UHFFFAOYSA-N
Synonym p-xylylenediamine, 1,4-phenylenedimethanamine, 1,4-bis aminomethyl benzene, 1,4-benzenedimethanamine, 1,4-xylylenediamine, p-xylylendiamine, 4-aminomethylbenzylamine, p-xylylendiamine czech, p-phenylenebis methylamine, 4-aminomethyl phenyl methanamine
PubChem CID 68315
IUPAC Name [4-(aminomethyl)phenyl]methanamine
SMILES C1=CC(=CC=C1CN)CN

Specifications

Melting Point 59°C to 63°C
Boiling Point 230°C (10 mmHg)
Flash Point 134°C (273°F)
Quantity 2 g
UN Number UN3259
Beilstein 2206190
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 136.2
Percent Purity 97%
Chemical Name or Material p-Xylylenediamine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber 208-719-3
RTECSNumber PF9000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only