missing translation for 'onlineSavingsMsg'
Learn More

p-Xylylene Dicyanide 98.0+%, TCI America™

Catalog No. X00615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00615G 5 g
X006125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X00615G Supplier TCI America Supplier No. X00615G

Chemical Identifiers

CAS 622-75-3
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00001923
InChI Key FUQCKESKNZBNOG-UHFFFAOYSA-N
Synonym 1,4-phenylenediacetonitrile, 1,4-benzenediacetonitrile, p-xylylene dicyanide, p-phenylenediacetonitrile, p-benzenediacetonitrile, 2,2'-1,4-phenylene diacetonitrile, p-bis cyanomethyl benzene, 1,4-bis cyanomethyl benzene, p-cyanomethyl benzyl cyanide, 2-4-cyanomethyl phenyl acetonitrile
PubChem CID 69327
IUPAC Name 2-[4-(cyanomethyl)phenyl]acetonitrile
SMILES C1=CC(=CC=C1CC#N)CC#N

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 156.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material p-Xylylene Dicyanide