missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Xylyl Cyanide 98.0+%, TCI America™
$57.79 - $413.83
Chemical Identifiers
| CAS | 2947-61-7 |
|---|---|
| Molecular Formula | C9H9N |
| Molecular Weight (g/mol) | 131.18 |
| MDL Number | MFCD00001922 |
| InChI Key | RNHKXHKUKJXLAU-UHFFFAOYSA-N |
| Synonym | 4-methylbenzyl cyanide, 4-methylphenylacetonitrile, p-tolylacetonitrile, p-methylbenzyl cyanide, 2-4-methylphenyl acetonitrile, benzeneacetonitrile, 4-methyl, 4-methylphenyl acetonitrile, 4-methylbenzeneacetonitrile, 4-methylbenzylcyanide, 2-p-tolylacetonitrile |
| PubChem CID | 76280 |
| IUPAC Name | 2-(4-methylphenyl)acetonitrile |
| SMILES | CC1=CC=C(CC#N)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
X004725ML
|
TCI America
X004725ML |
25 mL |
Each for $57.79
|
|
|||||
|
X0047500ML
|
TCI America
X0047500ML |
500 mL |
Each for $413.83
|
|
|||||
Chemical Identifiers
| 2947-61-7 | |
| 131.18 | |
| RNHKXHKUKJXLAU-UHFFFAOYSA-N | |
| 76280 | |
| CC1=CC=C(CC#N)C=C1 |
| C9H9N | |
| MFCD00001922 | |
| 4-methylbenzyl cyanide, 4-methylphenylacetonitrile, p-tolylacetonitrile, p-methylbenzyl cyanide, 2-4-methylphenyl acetonitrile, benzeneacetonitrile, 4-methyl, 4-methylphenyl acetonitrile, 4-methylbenzeneacetonitrile, 4-methylbenzylcyanide, 2-p-tolylacetonitrile | |
| 2-(4-methylphenyl)acetonitrile |
Specifications
| 2947-61-7 | |
| 245°C | |
| MFCD00001922 | |
| 3276 | |
| RNHKXHKUKJXLAU-UHFFFAOYSA-N | |
| 2-(4-methylphenyl)acetonitrile | |
| 76280 | |
| ≥98.0% (GC) | |
| p-Xylyl Cyanide |
| Yellow | |
| C9H9N | |
| 25 mL | |
| 4-methylbenzyl cyanide, 4-methylphenylacetonitrile, p-tolylacetonitrile, p-methylbenzyl cyanide, 2-4-methylphenyl acetonitrile, benzeneacetonitrile, 4-methyl, 4-methylphenyl acetonitrile, 4-methylbenzeneacetonitrile, 4-methylbenzylcyanide, 2-p-tolylacetonitrile | |
| CC1=CC=C(CC#N)C=C1 | |
| 131.18 | |
| 131.18 | |
| Liquid |
Safety and Handling
TSCA : No