Learn More
p-Xylene, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™
p-Xylene is an aromatic liquid, which is an isomeric form of xylene. It can be produced via Diels-Alder reaction between 2,5-dimethylfuran and ethylene catalyzed by zeolite.
Supplier:  Merck Emd Millipore 9568050ML
 View more versions of this product
Catalog No. 111018188
Only null left
Description
Description
Specifications
Specifications
Glass Bottle
9 mmHg (20°C)
0.861 g/mL (at 20°C)
C6H4(CH3)2
12°C to 13°C
1,4-Dimethylbenzene
106.17
Analytical Standard
106-42-3
3.7 (vs. air)
Neat
MFCD00008556
138°C
50 mL
≥99.5% (GC)
Product Suggestions
Product Suggestions