missing translation for 'onlineSavingsMsg'
Learn More

p-Xylene-2-sulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7126370
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06835G 5 g
D068325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D06835G Supplier TCI America Supplier No. D06835G
Only null left

Chemical Identifiers

CAS 19040-62-1
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.668
MDL Number MFCD00024875
InChI Key FZVZUIBYLZZOEW-UHFFFAOYSA-N
Synonym p-xylene-2-sulfonyl chloride, benzenesulfonyl chloride, 2,5-dimethyl, 2,5-dimethylbenzene-1-sulfonyl chloride, 2,5-dimethylphenylsulfonyl chloride, 2,5-dimethyl-benzenesulfonyl chloride, benzenesulfonylchloride, 2,5-dimethyl, 2,5-dimethylphenyl chlorosulfone, acmc-209etq, xylenesulfonylchloride, ksc178m0f
PubChem CID 87910
IUPAC Name 2,5-dimethylbenzenesulfonyl chloride
SMILES CC1=CC(=C(C=C1)C)S(=O)(=O)Cl

Specifications

Color White-Yellow
Boiling Point 150°C
Quantity 5 g
Physical Form Crystal at 20°C
UN Number 3261
Formula Weight 204.67
Percent Purity ≥98.0% (T)
Chemical Name or Material p-Xylene-2-sulfonyl Chloride