missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Xylene-2-sulfonyl Chloride 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 5 g |
| Physical Form | Crystal at 20°C |
| UN Number | 3261 |
| Boiling Point | 150°C |
| Formula Weight | 204.67 |
| Percent Purity | ≥98.0% (T) |
| Grade | Not Available |
| Chemical Name or Material | p-Xylene-2-sulfonyl Chloride |
Safety and Handling
| TSCA | Yes |