missing translation for 'onlineSavingsMsg'
Learn More

p-Xylene-2-sulfonic Acid Hydrate 98.0+%, TCI America™

Catalog No. D068125G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D068125G 25 g
D0681100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D068125G Supplier TCI America Supplier No. D068125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 609-54-1 | C8H10O3S | 186.23 g/mol

Chemical Identifiers

CAS 609-54-1
Molecular Formula C8H10O3S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00151010
InChI Key IRLYGRLEBKCYPY-UHFFFAOYSA-N
Synonym p-xylene-2-sulfonic acid, unii-l1sea5qnp9, p-xylene-2-sulphonic acid, benzenesulfonic acid, 2,5-dimethyl, l1sea5qnp9, 2,4-xylenesulfonic acid hydrate, 2,5-dimethyl-benzenesulfonic acid, 2,5-xylenesulfonic acid, p-xylenesulfonic acid, dsstox_cid_21641
PubChem CID 11868
IUPAC Name 2,5-dimethylbenzene-1-sulfonic acid
SMILES CC1=CC=C(C)C(=C1)S(O)(=O)=O

Specifications

Melting Point 86°C
Color Red-White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 186.23
Percent Purity ≥98.0% (HPLC,T)
Grade Not Available
Chemical Name or Material p-Xylene-2-sulfonic Acid Hydrate
EINECSNumber (9)-2093
TSCA Yes