missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyl Isovalerate 98.0+%, TCI America™

Catalog No. I039725ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I039725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I039725ML Supplier TCI America Supplier No. I039725ML
Only null left

Chemical Identifiers

CAS 55066-56-3
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00036648
InChI Key MVDPTWHTUYDLTL-UHFFFAOYSA-N
Synonym Isovaleric Acid p-Tolyl Ester
PubChem CID 62092
IUPAC Name (4-methylphenyl) 3-methylbutanoate
SMILES CC1=CC=C(C=C1)OC(=O)CC(C)C

Specifications

Color Yellow
Boiling Point 125°C
Quantity 25 mL
Formula Weight 192.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material p-Tolyl Isovalerate