missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyl Isothiocyanate 98.0+%, TCI America™

Catalog No. p-7131381
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03935G 5 g
I039325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03935G Supplier TCI America Supplier No. I03935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 622-59-3
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
MDL Number MFCD00004813
InChI Key ABQKHKWXTUVKGF-UHFFFAOYSA-N
Synonym p-tolyl isothiocyanate, 4-methylphenyl isothiocyanate, 4-methylphenylisothiocyanate, p-tolylisothiocyanate, 4-tolylisothiocyanate, benzene, 1-isothiocyanato-4-methyl, 4-tolyl isothiocyanate, isothiocyanic acid, p-tolyl ester, 1-isothiocyanato-4-methyl-benzene, isothiocyanic acid, 4-tolyl ester
PubChem CID 12149
IUPAC Name 1-isothiocyanato-4-methylbenzene
SMILES CC1=CC=C(C=C1)N=C=S

Specifications

Color White-Yellow
Boiling Point 237°C
Quantity 5 g
UN Number 2811
Formula Weight 149.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material p-Tolyl Isothiocyanate
RTECSNumber NX9500000
TSCA No