missing translation for 'onlineSavingsMsg'
Learn More

p-Toluoylacetonitrile, 97%

CAS: 7391-28-8 | C10H9NO | 159.19 g/mol

$49.26 - $591.38

Chemical Identifiers

CAS 7391-28-8
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00067895
InChI Key AIECDYDQPCANJK-UHFFFAOYSA-N
Synonym 4-methylbenzoylacetonitrile, 4-toluoylacetonitrile, 3-4-methylphenyl-3-oxopropanenitrile, p-toluoylacetonitrile, 3-oxo-3-p-tolyl propanenitrile, 3-oxo-3-p-tolylpropanenitrile, acmc-20amdj, pubchem12072, maybridge1_004714, 4-methyl-benzoylacetonitrile
PubChem CID 522513
IUPAC Name 3-(4-methylphenyl)-3-oxopropanenitrile
SMILES CC1=CC=C(C=C1)C(=O)CC#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1095003
View Documents
Thermo Scientific Chemicals
A1095003
1 g
Each for $49.26
Only null left
 
AAA1095006
View Documents
Thermo Scientific Chemicals
A1095006
5 g
Each for $168.95
Only null left
 
AAA1095014
View Documents
Thermo Scientific Chemicals
A1095014
25 g
Each for $591.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7391-28-8
159.19
AIECDYDQPCANJK-UHFFFAOYSA-N
522513
CC1=CC=C(C=C1)C(=O)CC#N
C10H9NO
MFCD00067895
4-methylbenzoylacetonitrile, 4-toluoylacetonitrile, 3-4-methylphenyl-3-oxopropanenitrile, p-toluoylacetonitrile, 3-oxo-3-p-tolyl propanenitrile, 3-oxo-3-p-tolylpropanenitrile, acmc-20amdj, pubchem12072, maybridge1_004714, 4-methyl-benzoylacetonitrile
3-(4-methylphenyl)-3-oxopropanenitrile

Specifications

7391-28-8
C10H9NO
1 g
4-methylbenzoylacetonitrile, 4-toluoylacetonitrile, 3-4-methylphenyl-3-oxopropanenitrile, p-toluoylacetonitrile, 3-oxo-3-p-tolyl propanenitrile, 3-oxo-3-p-tolylpropanenitrile, acmc-20amdj, pubchem12072, maybridge1_004714, 4-methyl-benzoylacetonitrile
CC1=CC=C(C=C1)C(=O)CC#N
159.19
159.19
p-Toluoylacetonitrile
99°C to 104°C
MFCD00067895
742923
AIECDYDQPCANJK-UHFFFAOYSA-N
3-(4-methylphenyl)-3-oxopropanenitrile
522513
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only