missing translation for 'onlineSavingsMsg'
Learn More

p-Toluoylacetonitrile, 97%

Catalog No. AAA1095003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1095003 1 g
AAA1095006 5 g
AAA1095014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1095003 Supplier Thermo Scientific Chemicals Supplier No. A1095003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7391-28-8 | C10H9NO | 159.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7391-28-8
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00067895
InChI Key AIECDYDQPCANJK-UHFFFAOYSA-N
Synonym 4-methylbenzoylacetonitrile, 4-toluoylacetonitrile, 3-4-methylphenyl-3-oxopropanenitrile, p-toluoylacetonitrile, 3-oxo-3-p-tolyl propanenitrile, 3-oxo-3-p-tolylpropanenitrile, acmc-20amdj, pubchem12072, maybridge1_004714, 4-methyl-benzoylacetonitrile
PubChem CID 522513
IUPAC Name 3-(4-methylphenyl)-3-oxopropanenitrile
SMILES CC1=CC=C(C=C1)C(=O)CC#N

Specifications

Melting Point 99°C to 104°C
Quantity 1 g
Beilstein 742923
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material p-Toluoylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only