Learn More
p-Tolunitrile, 98+%
CAS: 104-85-8 | C8H7N | 117.15 g/mol
$120.64 - $120.64
Chemical Identifiers
| CAS | 104-85-8 |
|---|---|
| Molecular Formula | C8H7N |
| Molecular Weight (g/mol) | 117.15 |
| MDL Number | MFCD00001827 |
| InChI Key | VCZNNAKNUVJVGX-UHFFFAOYSA-N |
| Synonym | p-tolunitrile, 4-cyanotoluene, benzonitrile, 4-methyl, 4-tolunitrile, p-methylbenzonitrile, p-toluonitrile, p-cyanotoluene, p-tolylnitrile, p-tolunitril, p-toluenenitrile |
| PubChem CID | 7724 |
| IUPAC Name | 4-methylbenzonitrile |
| SMILES | CC1=CC=C(C=C1)C#N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC158021000
|
Thermo Scientific Chemicals
158021000 |
100 g | Glass bottle |
Each for $120.64
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 104-85-8 | |
| 117.15 | |
| VCZNNAKNUVJVGX-UHFFFAOYSA-N | |
| 7724 | |
| CC1=CC=C(C=C1)C#N |
| C8H7N | |
| MFCD00001827 | |
| p-tolunitrile, 4-cyanotoluene, benzonitrile, 4-methyl, 4-tolunitrile, p-methylbenzonitrile, p-toluonitrile, p-cyanotoluene, p-tolylnitrile, p-tolunitril, p-toluenenitrile | |
| 4-methylbenzonitrile |
Specifications
| 104-85-8 | |
| 0.9800g/mL | |
| 85°C | |
| 98% min. (GC) | |
| C8H7N | |
| CH3C6H4CN | |
| 100 g | |
| 0.98 | |
| p-tolunitrile, 4-cyanotoluene, benzonitrile, 4-methyl, 4-tolunitrile, p-methylbenzonitrile, p-toluonitrile, p-cyanotoluene, p-tolylnitrile, p-tolunitril, p-toluenenitrile | |
| VCZNNAKNUVJVGX-UHFFFAOYSA-N | |
| 4-methylbenzonitrile | |
| 7724 | |
| 98+% | |
| p-Tolunitrile, 98% |
| 26.0°C to 28.0°C | |
| 103.0°C to 106.0°C (20.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.5285 to 1.5305 | |
| MFCD00001827 | |
| 09, 489 | |
| 15, 9698 | |
| Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
| CC1=CC=C(C=C1)C#N | |
| 117.15 | |
| 117.15 | |
| Viscous Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 203-244-8
RTECSNumber : XV0700000
TSCA : TSCA
RUO – Research Use Only