missing translation for 'onlineSavingsMsg'
Learn More

p-Tolunitrile, 98+%

Catalog No. AC158021000
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158021000 100 g Glass bottle
AC158020050 5 g Glass bottle
AC158022500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC158021000 Supplier Thermo Scientific Chemicals Supplier No. 158021000
Only null left

CAS: 104-85-8 | C8H7N | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-85-8
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.15
MDL Number MFCD00001827
InChI Key VCZNNAKNUVJVGX-UHFFFAOYSA-N
Synonym p-tolunitrile, 4-cyanotoluene, benzonitrile, 4-methyl, 4-tolunitrile, p-methylbenzonitrile, p-toluonitrile, p-cyanotoluene, p-tolylnitrile, p-tolunitril, p-toluenenitrile
PubChem CID 7724
IUPAC Name 4-methylbenzonitrile
SMILES CC1=CC=C(C=C1)C#N

Specifications

Melting Point 26.0°C to 28.0°C
Density 0.9800g/mL
Boiling Point 103.0°C to 106.0°C (20.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5285 to 1.5305
Linear Formula CH3C6H4CN
Quantity 100 g
Beilstein 09, 489
Specific Gravity 0.98
Merck Index 15, 9698
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 117.15
Percent Purity 98+%
Physical Form Viscous Liquid
Chemical Name or Material p-Tolunitrile, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
EINECSNumber 203-244-8
RTECSNumber XV0700000
TSCA TSCA

RUO – Research Use Only