missing translation for 'onlineSavingsMsg'
Learn More

p-Toluidine 99.0+%, TCI America™

Catalog No. T0300500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0300500G 500 g
T030025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0300500G Supplier TCI America Supplier No. T0300500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-49-0
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00007906
InChI Key RZXMPPFPUUCRFN-UHFFFAOYSA-N
Synonym p-toluidine, 4-toluidine, p-tolylamine, 4-aminotoluene, benzenamine, 4-methyl, 4-methylbenzenamine, p-methylaniline, p-methylbenzenamine, p-toluidin, p-aminotoluene
PubChem CID 7813
ChEBI CHEBI:37825
IUPAC Name 4-methylaniline
SMILES CC1=CC=C(C=C1)N

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 201°C
Quantity 500 g
UN Number 3451
Formula Weight 107.16
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material p-Toluidine
EINECSNumber (3)-0186
RTECSNumber XU3150000
TSCA Yes
Recommended Storage Refrigerator