missing translation for 'onlineSavingsMsg'
Learn More

p-Toluidine, For Spectrophotometric Det. of Au, Tl(III), W, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. p-7187739
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
Catalog No. Quantity
111019343 100 g
111018202 500 g
2 options

Catalog No. 111019343

Supplier: Merck Emd Millipore 89630100G

Only null left

Chemical Identifiers

MDL Number MFCD00007906
Synonym 4-Aminotoluene; 4-Methylaniline

Specifications

Melting Point 41°C to 46°C; 42°C to 45°C
Density 0.973 g/mL (at 25°C)
CAS 106-49-0
Boiling Point 200°C
Flash Point 87°C - Closed Cup
Ignition Point 899°F
Linear Formula CH3C6H4 NH2
Quantity 100 g
Solubility Information Ethanol: 1 g/10 mL (Hot)
Vapor Density 3.9 (vs. air)
Vapor Pressure 7 hPa (68°C)
Formula Weight 107.15
Percent Purity ≥99.0% (GC); ≥99.0%
Show More Show Less
RTECSNumber XU3150000