missing translation for 'onlineSavingsMsg'
Learn More

p-Toluidine-2-sulfonic Acid 98.0+%, TCI America™

Catalog No. A032825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A032825G 25 g
A0328500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A032825G Supplier TCI America Supplier No. A032825G

Chemical Identifiers

CAS 88-44-8
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.21
MDL Number MFCD00007908
InChI Key LTPSRQRIPCVMKQ-UHFFFAOYSA-N
Synonym 4-aminotoluene-3-sulfonic acid, ptmsptmsa, ptms, ptmsa, benzenesulfonic acid, 2-amino-5-methyl, 6-amino-m-toluenesulfonic acid, red 4b acid, p-toluidine-2-sulfonic acid, p-toluidine-m-sulfonic acid, m-toluenesulfonic acid, 6-amino
PubChem CID 6934
IUPAC Name 2-amino-5-methylbenzene-1-sulfonic acid
SMILES CC1=CC=C(N)C(=C1)S(O)(=O)=O

Specifications

Quantity 25 g
Formula Weight 187.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material p-Toluidine-2-sulfonic Acid
EINECSNumber (3)-1978
RTECSNumber XT6320000
TSCA Yes