missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonyl Isocyanate 95.0+%, TCI America™

Catalog No. T099825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T099825G 25 g
T0998100G 100 g
T0998500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T099825G Supplier TCI America Supplier No. T099825G

Chemical Identifiers

CAS 4083-64-1
Molecular Formula C8H7NO3S
Molecular Weight (g/mol) 197.21
MDL Number MFCD00002030
InChI Key VLJQDHDVZJXNQL-UHFFFAOYSA-N
Synonym tosyl isocyanate, p-toluenesulfonyl isocyanate, 4-methylbenzenesulfonyl isocyanate, p-toluenesulphonyl isocyanate, benzenesulfonyl isocyanate, 4-methyl, tosylisocyanate, p-tolylsulfonyl isocyanate, unii-h9004fjx6v, p-toluenesulfonic acid, anhydride with isocyanic acid, p-toluene sulfonyl isocyanate
PubChem CID 77703
IUPAC Name 4-methylbenzene-1-sulfonyl isocyanate
SMILES CC1=CC=C(C=C1)S(=O)(=O)N=C=O

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 130°C
Quantity 25 g
Formula Weight 197.21
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material p-Toluenesulfonyl Isocyanate
EINECSNumber (3)-2222
RTECSNumber DB9032000
TSCA Yes